3-(Phenylsulfonyl)propionitrile 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20196-06
L20196-14
CAAAAL20196-06EA
83.58
CAD
CAAAAL20196-06
CAAAAL20196-14
3-(Phenylsulfonyl)propionitrile 99%
3-(Phenylsulfonyl)propionitrile
Purity: 99%
CAS No.: 10154-75-3
Molecular Formula: C9H9NO2S
Formula Weight: 195.24
Odor: Odorless
Melting Point: 94-96°
Beilstein Reference: 2051019
MDL No.: MFCD00047798
Formula:
C₉H₉NO₂S MW: 195.24 g/mol Melting Pt: 94…96 °C Storage Temperature: Ambient |
MDL Number:
MFCD00047798 CAS Number: 10154-75-3 EINECS: 233-424-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...