2-Nitrophenyl selenocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L20193-03 L20193-06
CAAAAL20193-03EA 126.24 CAD
CAAAAL20193-03 CAAAAL20193-06
2-Nitrophenyl selenocyanate 97%
2-Nitrophenyl selenocyanate

Purity: 97% CAS No.: 51694-22-5 Molecular Formula: C7H4N2O2Se Formula Weight: 227.08 Melting Point: 134-141° Beilstein Reference: 1309777 UN No.: 3283 MDL No.: MFCD00043146

Formula: C₇H₄N₂O₂Se
MW: 227.092 g/mol
Melting Pt: 134…141 °C
Storage Temperature: Ambient
MDL Number: MFCD00043146
CAS Number: 51694-22-5
EINECS: 257-350-4
UN: 3283
ADR: 6.1 III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR