Methyl-2-amino-5-chlorobenzoate 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20187-06
L20187-14
CAAAAL20187-06EA
133.19
CAD
CAAAAL20187-06
CAAAAL20187-14
Methyl-2-amino-5-chlorobenzoate 98+%
Methyl 2-amino-5-chlorobenzoate
Purity: 98+%
CAS No.: 5202-89-1
Molecular Formula: C8H8ClNO2
Formula Weight: 185.61
Melting Point: 67-70°
Boiling Point: 168-170°/1mm
MDL No.: MFCD00007837
Formula:
C₈H₈ClNO₂ Boiling Pt: 168…170 °C/1 mm Melting Pt: 67…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007837 CAS Number: 5202-89-1 EINECS: 225-992-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...