4-(Dimethylamino)piperidine 97%

Supplier: Thermo Scientific Chemicals
Danger

L20176-06 L20176-14
CAAAAL20176-06EA 230.68 CAD
CAAAAL20176-06 CAAAAL20176-14
4-(Dimethylamino)piperidine 97%
4-(Dimethylamino)piperidine

Purity: 97% CAS No.: 50533-97-6 Molecular Formula: C7H16N2 Formula Weight: 128.22 Refractive Index: 1.4760 Boiling Point: 187° Beilstein Reference: 1190 UN No.: 2735 MDL No.: MFCD00023144

Formula: C₇H₁₆N₂
Boiling Pt: 187 °C
Storage Temperature: Ambient
MDL Number: MFCD00023144
CAS Number: 50533-97-6
EINECS: 256-617-2
UN: 2735
ADR: "8",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR