(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide 98%

Supplier: Thermo Scientific Chemicals
Warning

L20170-03 L20170-MD
CAAAAL20170-03EA 303.36 CAD
CAAAAL20170-03 CAAAAL20170-MD
(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide 98%
(1S,4S)-2,5-Diazabicyclo[2.2.1]heptane dihydrobromide

Purity: 98% CAS No.: 132747-20-7 Molecular Formula: C5H10N2.2HBr Formula Weight: 259.97 Melting Point: ca 290° dec. Beilstein Reference: 4258260 Optical Rotation: +22° (c=1 in water) MDL No.: MFCD08272741

Formula: C₅H₁₀N₂
MW: 259.97 g/mol
Melting Pt: ∼ 290 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01321289
CAS Number: 132747-20-7

Order Now

Specification Test Results

Optical rotation +22° (c=1 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR