(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20155-03
L20155-06
L20155-14
CAAAAL20155-03EA
129.09
CAD
CAAAAL20155-03
CAAAAL20155-06
CAAAAL20155-14
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt 96%
(Methoxycarbonylsulfamoyl)triethylammonium hydroxide inner salt
Fieser: 14,208 21,266
Purity: 96%
CAS No.: 29684-56-8
Molecular Formula: C8H18N2O4S
Formula Weight: 238.30
Storage Temperature: -30°C to -10°C
Melting Point: 72-82°
Beilstein Reference: 1432131
MDL No.: MFCD00077815
Formula:
C₈H₁₈N₂O₄S Melting Pt: 72…82 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00077815 CAS Number: 29684-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...