(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
tert-butyl (1S,4S)-2,5-diazabicyclo[2.2.1]heptane-2-carboxylate
L20131-03
L20131-06
CAAAAL20131-03EA
165.9
CAD
CAAAAL20131-03
CAAAAL20131-06
CAAAAL20131-MD
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane 97%
(1S,4S)-2-Boc-2,5-diazabicyclo[2.2.1]heptane
Purity: 97%
CAS No.: 113451-59-5
Molecular Formula: C10H18N2O2
Formula Weight: 198.26
Storage Temperature: 2°C to 8°C
Melting Point: 74-76°
Beilstein Reference: 6460311
Optical Rotation: -44° (c=1 in chloroform)
MDL No.: MFCD01569250
Formula:
C₁₀H₁₈N₂O₂ Melting Pt: 74…76 °C Storage Temperature: Ambient |
MDL Number:
MFCD01569250 CAS Number: 113451-59-5 |
Specification Test Results
Optical rotation | -44° (c=1 in chloroform) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...