trans-Bis(tributylstannyl)ethylene 96%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: trans-1,2-Bis(tri-n-butylstannyl)ethylene

L20130-03 L20130-06
CAAAAL20130-03EA 214.88 CAD
CAAAAL20130-03 CAAAAL20130-06
trans-Bis(tributylstannyl)ethylene 96%
trans-Bis(tributylstannyl)ethylene

Purity: 96% CAS No.: 14275-61-7 Molecular Formula: C26H56Sn2 Formula Weight: 606.15 Storage Temperature: 2°C to 8°C Refractive Index: 1.5020 Boiling Point: 185°/0.01mm Beilstein Reference: 3613803 UN No.: 2788 MDL No.: MFCD01631299

Formula: C₂₆H₅₆Sn₂
MW: 606.15
Boiling Pt: 185 °C/0.01 mm
Density: 1.140
Storage Temperature: Ambient
MDL Number: MFCD01631299
CAS Number: 14275-61-7
UN: 2788
ADR: "6.1",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR