3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 99%
Supplier: Thermo Scientific Chemicals
L20129-06
L20129-MD
CAAAAL20129-06EA
496.12
CAD
CAAAAL20129-06
CAAAAL20129-MD
3H-1,2-Benzodithiol-one-1,1-dioxide (beaucage reagent) 99%
3H-1,2-Benzodithiol-one 1,1-dioxide (Beaucage Reagent)
Purity: 99%
CAS No.: 66304-01-6
Molecular Formula: C7H4O3S2
Formula Weight: 200.24
Melting Point: 102-107°
Beilstein Reference: 4180332
UN No.: 3335
MDL No.: MFCD00132960
Formula:
C₇H₄O₃S₂ Melting Pt: 102…107 °C Storage Temperature: Ambient |
MDL Number:
MFCD00132960 CAS Number: 66304-01-6 UN: 3335 ADR: 9,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...