2-(Benzyloxy)phenylboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Benzyloxyphenylboronic acid, (2-(Benzyloxy)phenyl)boronic acid, 2-Benzyloxybenzeneboronic acid

L20100-03 L20100-06
CAAAAL20100-03EA 73.15 CAD
CAAAAL20100-03 CAAAAL20100-06
2-(Benzyloxy)phenylboronic acid 96%
2-(Benzyloxy)phenylboronic acid

Purity: 96% CAS No.: 190661-29-1 Molecular Formula: C13H13BO3 Formula Weight: 228.06 Melting Point: 96-99° Beilstein Reference: 7813704

Formula: C₁₃H₁₃BO₃
MW: 228.06 g/mol
Melting Pt: 96…99 °C
Storage Temperature: Ambient
MDL Number: MFCD01632206
CAS Number: 190661-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR