2-Methoxy-5-pyridineboronic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20087-03
L20087-06
CAAAAL20087-03EA
109.34
CAD
CAAAAL20087-03
CAAAAL20087-06
CAAAAL20087-MD
2-Methoxy-5-pyridineboronic acid 95%
2-Methoxy-5-pyridineboronic acid
Purity: 95%
CAS No.: 163105-89-3
Molecular Formula: C6H8BNO3
Formula Weight: 152.95
Melting Point: 132-133°
Beilstein Reference: 9119950
Formula:
C₆H₈BNO₃ Melting Pt: 132…133 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093044 CAS Number: 163105-89-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...