2-Iodo-3-methylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L20047-03 L20047-06
CAAAAL20047-03EA 93.06 CAD
CAAAAL20047-03 CAAAAL20047-06
2-Iodo-3-methylpyridine 97%
2-Iodo-3-methylpyridine

Purity: 97% CAS No.: 22282-58-2 Molecular Formula: C6H6IN Formula Weight: 219.03 Storage Temperature: -30°C to -10°C Refractive Index: 1.6250 Boiling Point: 80°/0.3mm Beilstein Reference: 5328169 MDL No.: MFCD04039350

Formula: C₆H₆IN
Boiling Pt: 80 °C/0.3 mm
Storage Temperature: Refrigerator
MDL Number: MFCD04039350
CAS Number: 22282-58-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR