5-Iodopyridin-2-ol 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-5-iodopyridine, 5-Iodopyridin-2(1H)-one

L20044-03 L20044-06
CAAAAL20044-03EA 37.6 CAD
CAAAAL20044-03 CAAAAL20044-06
5-Iodopyridin-2-ol 97%
5-Iodopyridin-2-ol

Purity: 97% CAS No.: 13472-79-2 Molecular Formula: C5H4INO Formula Weight: 221.00 Melting Point: 189-191° Beilstein Reference: 108752 MDL No.: MFCD00234058

Formula: C₅H₄INO
MW: 221 g/mol
Melting Pt: 189…191 °C
Storage Temperature: Ambient
MDL Number: MFCD00234058
CAS Number: 13472-79-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR