1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20001-22
L20001-36
CAAAAL20001-22EA
54.98
CAD
CAAAAL20001-22
CAAAAL20001-36
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers) 97%
1,2-Cyclohexanedicarboxylic anhydride (mixture of cis and trans isomers)
Purity: 97%
CAS No.: 85-42-7
Molecular Formula: C8H10O3
Formula Weight: 154.17
Flash Point: 275°(527°F)
Melting Point: 32-34°
Boiling Point: 158°/17mm
Beilstein Reference: 83213
MDL No.: MFCD00064863
Formula:
C₈H₁₀O₃ Boiling Pt: 158 °C/17 mm Melting Pt: 32…34 °C Flash Pt: 275 °C(527 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00064863 CAS Number: 85-42-7 EINECS: 201-604-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...