4-Iodobenzylamine hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (4-Iodophenyl)methanamine hydrochloride

L19967-03 L19967-06
CAAAAL19967-03EA 122.13 CAD
CAAAAL19967-03 CAAAAL19967-06
4-Iodobenzylamine hydrochloride 97%
4-Iodobenzylamine hydrochloride

Purity: 97% CAS No.: 59528-27-7 Molecular Formula: C7H8IN.HCl Formula Weight: 269.51 Melting Point: 275-280° dec. Beilstein Reference: 3691299

Formula: C₇H₉ClIN
MW: 269.51 g/mol
Melting Pt: 275…280 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD04039324
CAS Number: 59528-27-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR