4-(Cyanomethyl)phenylboronic acid 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19955-03
L19955-06
CAAAAL19955-03EA
148.52
CAD
CAAAAL19955-03
CAAAAL19955-06
4-(Cyanomethyl)phenylboronic acid 98%
4-(Cyanomethyl)phenylboronic acid
Purity: 98%
CAS No.: 91983-26-5
Molecular Formula: C8H8BNO2
Formula Weight: 160.97
Melting Point: ca 196-200°
Beilstein Reference: 3031323
MDL No.: MFCD01632200
Formula:
C₈H₈BNO₂ Melting Pt: ca 196…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD01632200 CAS Number: 91983-26-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...