2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
4-(Bromomethyl)phenylboronic acid Pinacol ester, 4-Bromomethylphenylboronic acid, pinacol ester, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl bromide, 2-[4-(Bromomethyl)phenyl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
L19954-03
L19954-06
CAAAAL19954-03EA
197.5
CAD
CAAAAL19954-03
CAAAAL19954-06
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane 95%
2-(4-(Bromomethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Purity: 95%
CAS No.: 138500-85-3
Molecular Formula: C13H18BBrO2
Formula Weight: 297.00
Melting Point: 83-85°
Beilstein Reference: 7921808
UN No.: 3261
MDL No.: MFCD02179493
Formula:
C₁₃H₁₈BBrO₂ Melting Pt: 83…85 °C Storage Temperature: Ambient |
MDL Number:
MFCD02179493 CAS Number: 138500-85-3 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...