N-Boc-1,6-diaminohexane 98%

Supplier: Thermo Scientific Chemicals
Danger

L19948-03 L19948-06 L19948-14
CAAAAL19948-03EA 82.32 CAD
CAAAAL19948-03 CAAAAL19948-06 CAAAAL19948-14
N-Boc-1,6-diaminohexane 98%
N-Boc-1,6-diaminohexane

Purity: 98% CAS No.: 51857-17-1 Molecular Formula: (CH3)3COCONH(CH2)6NH2 Formula Weight: 216.33 Flash Point: 110°(230°F) Refractive Index: 1.4620 Boiling Point: 106-110°/0.3mm Beilstein Reference: 2089264 UN No.: 2735 MDL No.: MFCD00671489

Formula: C₁₁H₂₄N₂O₂
Boiling Pt: 106…110 °C/0.3 mm
Density: 0.965
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00671489
CAS Number: 51857-17-1
EINECS: 628-984-2
UN: 2735
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR