4-Fluoro-L-phenylalanine 98+%
Supplier: Thermo Scientific Chemicals
Synonyms:
(S)-2-Amino-3-(4-fluorophenyl)propanoic acid, H-Phe(4-F)-OH
L19934-03
L19934-06
L19934-MD
CAAAAL19934-03EA
338.12
CAD
CAAAAL19934-03
CAAAAL19934-06
CAAAAL19934-MD
4-Fluoro-L-phenylalanine 98+%
4-Fluoro-L-phenylalanine
Purity: 98+%
CAS No.: 1132-68-9
Molecular Formula: C9H10FNO2
Formula Weight: 183.18
Melting Point: ca 240° dec.
Beilstein Reference: 2416148
Formula:
C₉H₁₀FNO₂ Melting Pt: ca 240 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00063064 CAS Number: 1132-68-9 |
Specification Test Results
Optical rotation | -25° (c=1 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...