4'-Fluoro-3'-methoxyacetophenone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-(4-Fluoro-3-methoxyphenyl)ethanone
L19894-03
L19894-MD
CAAAAL19894-03EA
350.76
CAD
CAAAAL19894-03
CAAAAL19894-MD
4'-Fluoro-3'-methoxyacetophenone 98%
4'-Fluoro-3'-methoxyacetophenone
Purity: 98%
CAS No.: 64287-19-0
Molecular Formula: C9H9FO2
Formula Weight: 168.17
Melting Point: 51-53°
Beilstein Reference: 4381319
Formula:
C₉H₉FO₂ MW: 168.17 g/mol Melting Pt: 51…53 °C Storage Temperature: Ambient |
MDL Number:
MFCD00272135 CAS Number: 64287-19-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...