Methyl-2-aminoisonicotinate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19869-03
L19869-06
CAAAAL19869-03EA
38.71
CAD
CAAAAL19869-03
CAAAAL19869-06
Methyl-2-aminoisonicotinate 97%
Methyl 2-aminoisonicotinate
Purity: 97%
CAS No.: 6937-03-7
Molecular Formula: C7H8N2O2
Formula Weight: 152.15
Melting Point: 145-149°
Beilstein Reference: 128642
Formula:
C₇H₈N₂O₂ Melting Pt: 145…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD04039316 CAS Number: 6937-03-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...