2-Aminoisonicotinamide 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Aminopyridine-4-carboxamide, 2-Amino-4-pyridinecarboxamide

L19843-03 L19843-06 L19843-14
CAAAAL19843-03EA 79.63 CAD
CAAAAL19843-03 CAAAAL19843-06 CAAAAL19843-14
2-Aminoisonicotinamide 95%
2-Aminoisonicotinamide

Purity: 95% CAS No.: 13538-42-6 Molecular Formula: C6H7N3O Formula Weight: 137.14 Melting Point: 252-258° dec. Beilstein Reference: 115924

Formula: C₆H₇N₃O
MW: 137.14 g/mol
Melting Pt: 252…258 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03791262
CAS Number: 13538-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR