Dibenzothiophene-4-boronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Dibenzothiopheneboronic acid

L19831-03 L19831-06
CAAAAL19831-03EA 76.47 CAD
CAAAAL19831-03 CAAAAL19831-06
Dibenzothiophene-4-boronic acid 95%
Dibenzothiophene-4-boronic acid

Purity: 95% CAS No.: 108847-20-7 Molecular Formula: C12H9BO2S Formula Weight: 228.08 Melting Point: 300° Beilstein Reference: 197863

Formula: C₁₂H₉BO₂S
MW: 228.08 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD01318182
CAS Number: 108847-20-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR