4-Dibenzofuranboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dibenzofuran-4-boronic acid, Dibenzo[b,d]furan-4-ylboronic acid
L19830-03
L19830-06
CAAAAL19830-03EA
47.56
CAD
CAAAAL19830-03
CAAAAL19830-06
4-Dibenzofuranboronic acid 98+%
4-Dibenzofuranboronic acid
Purity: 98+%
CAS No.: 100124-06-9
Molecular Formula: C12H9BO3
Formula Weight: 212.01
Melting Point: 286-291°
Beilstein Reference: 1645426
Formula:
C₁₂H₉BO₃ MW: 212.01 g/mol Melting Pt: 286…291 °C Storage Temperature: Ambient |
MDL Number:
MFCD00092336 CAS Number: 100124-06-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...