Fmoc-1-aminocyclohexane-1-carboxylic acid ≥98%

Supplier: Thermo Scientific Chemicals

L19810-03 L19810-06
CAAAAL19810-03EA 120.4 CAD
CAAAAL19810-03 CAAAAL19810-06
Fmoc-1-aminocyclohexane-1-carboxylic acid ≥98%
Fmoc-1-aminocyclohexane-1-carboxylic acid

Purity: 98+% CAS No.: 162648-54-6 Molecular Formula: C22H23NO4 Formula Weight: 365.43 Storage Temperature: 2°C to 8°C Melting Point: 185-187° dec. Beilstein Reference: 7887646 MDL No.: MFCD00273464

Formula: C₂₂H₂₃NO₄
Melting Pt: 185…187 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00273464
CAS Number: 162648-54-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR