(±)-3-Amino-3-phenylpropionic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19801-03
L19801-06
CAAAAL19801-03EA
39.97
CAD
CAAAAL19801-03
CAAAAL19801-06
(±)-3-Amino-3-phenylpropionic acid 99%
(±)-3-Amino-3-phenylpropionic acid
Purity: 99%
CAS No.: 614-19-7
Molecular Formula: C9H11NO2
Formula Weight: 165.19
Storage Temperature: 2°C to 8°C
Melting Point: ca 220° dec.
Beilstein Reference: 2803286
MDL No.: MFCD00008064
Formula:
C₉H₁₁NO₂ Melting Pt: ∼ 220 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00008064 CAS Number: 614-19-7 EINECS: 210-371-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...