3-Amino-3-(3-chlorophenyl)propionic acid 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
3-Amino-3-(3-chlorophenyl)propanoic acid
L19797-03
L19797-MD
CAAAAL19797-03EA
115.18
CAD
CAAAAL19797-03
CAAAAL19797-MD
3-Amino-3-(3-chlorophenyl)propionic acid 98%
3-Amino-3-(3-chlorophenyl)propionic acid
Purity: 98%
CAS No.: 68208-21-9
Molecular Formula: C9H10ClNO2
Formula Weight: 199.64
Storage Temperature: 2°C to 8°C
Melting Point: ca 217° dec.
Beilstein Reference: 2938807
MDL No.: MFCD00463160
Formula:
C₉H₁₀ClNO₂ MW: 199.64 g/mol Melting Pt: ∼ 217 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00463160 CAS Number: 68208-21-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...