ω,ω-Dibromo-4-fluoroacetophenone 98%

Supplier: Thermo Scientific Chemicals
Danger

L19788-03 L19788-06
CAAAAL19788-03EA 20.54 CAD
CAAAAL19788-03 CAAAAL19788-06
ω,ω-Dibromo-4-fluoroacetophenone 98%
ω,ω-Dibromo-4-fluoroacetophenone

Purity: 98% CAS No.: 7542-64-5 Molecular Formula: C8H5Br2FO Formula Weight: 295.93 Storage Temperature: 2°C to 8°C Melting Point: 36-38° Beilstein Reference: 2256317 UN No.: 3261 MDL No.: MFCD03791309

Formula: C₈H₅Br₂FO
Melting Pt: 36…38 °C
Storage Temperature: Ambient
MDL Number: MFCD03791309
CAS Number: 7542-64-5
UN: 3261
ADR: 8,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR