Methyl-2-amino-5-fluorobenzoate 98%
Supplier: Thermo Scientific Chemicals
L19778-06
L19778-14
CAAAAL19778-06EA
88.8
CAD
CAAAAL19778-06
CAAAAL19778-14
Methyl-2-amino-5-fluorobenzoate 98%
Methyl 2-amino-5-fluorobenzoate
Purity: 98%
CAS No.: 319-24-4
Molecular Formula: C8H8FNO2
Formula Weight: 169.16
Flash Point: 132°(269°F)
Melting Point: 21-25°
Boiling Point: 100-102°/0.5mm
Beilstein Reference: 2803210
MDL No.: MFCD03791308
Formula:
C₈H₈FNO₂ Boiling Pt: 100…102 °C/0.5 mm Melting Pt: 21…25 °C Density: 1.260 Flash Pt: 132 °C(269 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03791308 CAS Number: 319-24-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...