Ethyl (R)-(-)-4-cyano-3-hydroxybutyate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(R)-4-Cyano-3-hydroxybutyric acid ethyl ester, BUTANOIC ACID, 4-CYANO-3-HYDROXY-, ETHYL ESTER, (3R)-, Ethyl (R)-4-cyano-3-hydroxybutanoate, (R)-ethyl 4-cyano-3-hydroxybutanoate
L19771-06
L19771-14
CAAAAL19771-06EA
81.69
CAD
CAAAAL19771-06
CAAAAL19771-14
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate 98%
Ethyl (R)-(-)-4-cyano-3-hydroxybutyate
Purity: 98%
CAS No.: 141942-85-0
Molecular Formula: C7H11NO3
Formula Weight: 157.17
Flash Point:
Formula:
C₇H₁₁NO₃ MW: 157.169 g/mol Boiling Pt: 270 °C Density: 1.114 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00270839 CAS Number: 141942-85-0 EINECS: 430-220-6 |
Specification Test Results
Optical rotation | -4.5° (neat) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...