Morpholinosulfur trifluoride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Morph-DAST
L19751-03
L19751-06
L19751-14
CAAAAL19751-03EA
64.62
CAD
CAAAAL19751-03
CAAAAL19751-06
CAAAAL19751-14
Morpholinosulfur trifluoride 95%
Morpholinosulfur trifluoride
Purity: 95%
CAS No.: 51010-74-3
Molecular Formula: C4H8F3NOS
Formula Weight: 175.17
Storage Temperature: -30°C to -10°C
Flash Point: 76°(168°F)
Refractive Index: 1.4545
Boiling Point: 41-42°/0.5mm
Beilstein Reference: 606224
UN No.: 3265
MDL No.: MFCD00037057
Formula:
C₄H₈F₃NOS MW: 175.17 g/mol Boiling Pt: 41…42 °C/0.5 mm Density: 1.436 Flash Pt: 76 °C (168 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00037057 CAS Number: 51010-74-3 UN: 3265 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...