Methyl-Β-L-arabinopyranoside 98

Supplier: Thermo Scientific Chemicals

Synonyms: (2S,3R,4S,5S)-2-Methoxytetrahydro-2H-pyran-3,4,5-triol

L19746-03 L19746-06
CAAAAL19746-03EA 103.17 CAD
CAAAAL19746-03 CAAAAL19746-06
Methyl-Β-L-arabinopyranoside 98
Methyl-Β-L-arabinopyranoside

Purity: 98+% CAS No.: 1825-00-9 Molecular Formula: C6H12O5 Formula Weight: 164.16 Melting Point: 167-172° Beilstein Reference: 4658388 Optical Rotation: +243° (c=2 in water) MDL No.: MFCD00047547

Formula: C₆H₁₂O₅
MW: 164.17 g/mol
Melting Pt: 167…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00047547
CAS Number: 1825-00-9
EINECS: 217-362-2

Order Now

Specification Test Results

Optical rotation +241° (c=2 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR