3-Acetoxy-1-propenylboronic acid pinacol ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
(E)-3-Acetoxy-1-propenylboronic acid pinacol ester, (E)-3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)allyl acetate
L19700-03
L19700-MD
CAAAAL19700-03EA
434.5
CAD
CAAAAL19700-03
CAAAAL19700-MD
3-Acetoxy-1-propenylboronic acid pinacol ester 97%
3-Acetoxy-1-propenylboronic acid pinacol ester
Purity: 97%
CAS No.: 161395-97-7
Molecular Formula: C11H19BO4
Formula Weight: 226.08
Refractive Index: 1.4485
Beilstein Reference: 6923397
MDL No.: MFCD03788751
Formula:
C₁₁H₁₉BO₄ MW: 226.08 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03788751 CAS Number: 161395-97-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...