4-Methyl-β-styrylboronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
(E)-4,4,5,5-Tetramethyl-2-(4-methylstyryl)-1,3,2-dioxaborolane
L19698-03
L19698-MD
CAAAAL19698-03EA
221.2
CAD
CAAAAL19698-03
CAAAAL19698-MD
4-Methyl-β-styrylboronic acid pinacol ester 98%
4-Methyl-beta-styrylboronic acid pinacol ester
Purity: 98%
CAS No.: 149777-84-4
Molecular Formula: C15H21BO2
Formula Weight: 244.14
Melting Point: 56-58°
Beilstein Reference: 7204616
MDL No.: MFCD03788749
Formula:
C₁₅H₂₁BO₂ MW: 244.14 g/mol Melting Pt: 56…58 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788749 CAS Number: 149777-84-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...