(R)-1-Methyl-3-pyrrolidinol 99%, ee 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19694-03
L19694-06
CAAAAL19694-03EA
127.35
CAD
CAAAAL19694-03
CAAAAL19694-06
CAAAAL19694-MD
(R)-1-Methyl-3-pyrrolidinol 99%, ee 99%
(R)-1-Methyl-3-pyrrolidinol
Purity: 99%
CAS No.: 104641-60-3
Molecular Formula: C5H11NO
Formula Weight: 101.15
Flash Point: 70°(158°F)
Refractive Index: 1.4640
Boiling Point: 103-106°/35mm
Beilstein Reference: 4349393
MDL No.: MFCD03788747
Formula:
C₅H₁₁NO Boiling Pt: 103…106 °C/35 mm Density: 0.920 Flash Pt: 70 °C(158 °F) Storage Temperature: Ambient |
MDL Number:
MFCD03788747 CAS Number: 104641-60-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...