(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%, ee 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: (R)-1-Benzylpyrrolidin-3-amine, (R)-(-)-1-Benzyl-3-aminopyrrolidine

L19690-03 L19690-06
CAAAAL19690-03EA 84.85 CAD
CAAAAL19690-03 CAAAAL19690-06
(3R)-(-)-1-Benzyl-3-aminopyrrolidine 99%, ee 99%
(3R)-(-)-1-Benzyl-3-aminopyrrolidine

Purity: 99% CAS No.: 114715-39-8 Molecular Formula: C11H16N2 Formula Weight: 176.26 Flash Point: 140°(284°F) Refractive Index: 1.5430 Boiling Point: 101-103°/3mm Beilstein Reference: 6477592 Optical Rotation: -2° (neat) UN No.: 2735 MDL No.: MFCD00082638

Formula: C₁₁H₁₆N₂
Boiling Pt: 101…103 °C/3 mm
Density: 1.020
Flash Pt: 140 °C(284 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082638
CAS Number: 114715-39-8
UN: 2735
ADR: "8",III

Order Now

Specification Test Results

Optical rotation -2° (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR