Hafnium(IV) trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19688-06
L19688-14
CAAAAL19688-06EA
172.22
CAD
CAAAAL19688-06
CAAAAL19688-14
Hafnium(IV) trifluoromethanesulfonate 98%
Hafnium(IV) trifluoromethanesulfonate
Purity: 98%
CAS No.: 161337-67-3
Molecular Formula: C4F12HfO12S4
Formula Weight: 774.75
Melting Point:
Formula:
CHF₃O₃S Melting Pt: >350 °C Storage Temperature: Ambient |
MDL Number:
MFCD01321255 CAS Number: 161337-67-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...