Copper(I) trifluoromethanesulfonate toluene complex (2:1) 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19656-03
L19656-06
CAAAAL19656-03EA
140.78
CAD
CAAAAL19656-03
CAAAAL19656-06
Copper(I) trifluoromethanesulfonate toluene complex (2:1) 98%
Copper(I) trifluoromethanesulfonate toluene complex
Purity: 98%
CAS No.: 48209-28-5
Molecular Formula: [CF3SO3Cu]2.C6H5CH3
Formula Weight: 517.37
Flash Point: 76°(168°F)
Melting Point: 180° dec.
UN No.: 1325
MDL No.: MFCD01863766
Formula:
[CF₃SO₃Cu]₂·C₆H₅CH₃ Melting Pt: 180 °C (dec.) Flash Pt: 76 °C (168 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01863766 CAS Number: 48209-28-5 UN: 1325 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...