1H-Pyrazole-4-boronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19654-03
L19654-06
CAAAAL19654-03EA
120.24
CAD
CAAAAL19654-03
CAAAAL19654-06
1H-Pyrazole-4-boronic acid pinacol ester 98%
1H-Pyrazole-4-boronic acid pinacol ester
Purity: 98%
CAS No.: 269410-08-4
Molecular Formula: C9H15BN2O2
Formula Weight: 194.04
Melting Point: 145-149°
MDL No.: MFCD03453063
Formula:
C₉H₁₅BN₂O₂ Melting Pt: 145…149 °C Storage Temperature: Ambient |
MDL Number:
MFCD03453063 CAS Number: 269410-08-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...