5-Isoquinolinecarboxylic acid 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19637-03
L19637-MD
CAAAAL19637-03EA
415.54
CAD
CAAAAL19637-03
CAAAAL19637-MD
5-Isoquinolinecarboxylic acid 96%
5-Isoquinolinecarboxylic acid
Purity: 96%
CAS No.: 27810-64-6
Molecular Formula: C10H7NO2
Formula Weight: 173.17
Melting Point: 278-281°
Boiling Point: 60-62°/11mm
MDL No.: MFCD03788744
Formula:
C₁₀H₇NO₂ Boiling Pt: 60…62 °C/11 mm Melting Pt: 278…281 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788744 CAS Number: 27810-64-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...