Isoquinoline-5-carbaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19636-03
L19636-MD
CAAAAL19636-03EA
138.88
CAD
CAAAAL19636-03
CAAAAL19636-MD
Isoquinoline-5-carbaldehyde 97%
Isoquinoline-5-carbaldehyde
Purity: 97%
CAS No.: 80278-67-7
Molecular Formula: C10H7NO
Formula Weight: 157.17
Melting Point: 113-116°
Beilstein Reference: 112875
Formula:
C₁₀H₇NO Melting Pt: 113…116 °C Storage Temperature: Ambient |
MDL Number:
MFCD03412483 CAS Number: 80278-67-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...