Benzothiazole-2-carbonyl chloride 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19631-03
L19631-MD
CAAAAL19631-03EA
225.94
CAD
CAAAAL19631-03
CAAAAL19631-MD
Benzothiazole-2-carbonyl chloride 95%
Benzothiazole-2-carbonyl chloride
Purity: 95%
CAS No.: 67748-61-2
Molecular Formula: C8H4ClNOS
Formula Weight: 197.64
Melting Point: 113-115°
Beilstein Reference: 130990
UN No.: 3261
MDL No.: MFCD03659697
Formula:
C₈H₄ClNOS Melting Pt: 113…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD03659697 CAS Number: 67748-61-2 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...