3-Bromobenzaldehyde ethylene acetal 98+%

Supplier: Thermo Scientific Chemicals

L19611-06 L19611-14
CAAAAL19611-06EA 61.3 CAD
CAAAAL19611-06 CAAAAL19611-14
3-Bromobenzaldehyde ethylene acetal 98+%
3-Bromobenzaldehyde ethylene acetal

Purity: 98+% CAS No.: 17789-14-9 Molecular Formula: C9H9BrO2 Formula Weight: 229.07 Flash Point: 110°(230°F) Refractive Index: 1.5630 Boiling Point: 132-133°/8mm Beilstein Reference: 1344030 MDL No.: MFCD00003209

Formula: C₉H₉BrO₂
Boiling Pt: 132…133 °C/8 mm
Density: 1.514
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00003209
CAS Number: 17789-14-9
EINECS: 241-766-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR