2-(9-Carbazolyl)ethylboronic acid pinacol ester 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
9-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)ethyl)-9H-carbazole
L19580-03
L19580-MD
CAAAAL19580-03EA
284.4
CAD
CAAAAL19580-03
CAAAAL19580-MD
2-(9-Carbazolyl)ethylboronic acid pinacol ester 98%
2-(9-Carbazolyl)ethylboronic acid pinacol ester
Purity: 98%
CAS No.: 608534-41-4
Molecular Formula: C20H24BNO2
Formula Weight: 321.23
Melting Point: 111-113°
MDL No.: MFCD03788738
Formula:
C₂₀H₂₄BNO₂ MW: 321.23 g/mol Melting Pt: 111…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788738 CAS Number: 608534-41-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...