Amberlite® IRC-748, Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

Synonyms: Amberlite® IRC-748 IRC-748, Amberlite® IRC-748

L19570-0B L19570-30
CAAAAL19570-0BEA 233.84 CAD
CAAAAL19570-0B CAAAAL19570-30
Amberlite® IRC-748, Amberlite™ ion exchange resin
Amberlite® IRC-748

CAS No.: 79620-28-3 Odor: Odorless MDL No.: MFCD00132702

Storage Temperature: Ambient
MDL Number: MFCD00132702
CAS Number: 79620-28-3
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR