Amberlite® IRA-900 (Cl), Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

L19567-0B L19567-30
CAAAAL19567-0BEA 167.48 CAD
CAAAAL19567-0B CAAAAL19567-30
Amberlite® IRA-900 (Cl), Amberlite™ ion exchange resin
Amberlite® IRA-900 (Cl)

CAS No.: 9050-97-9 Odor: Odorless MDL No.: MFCD00132712

Storage Temperature: Ambient
MDL Number: MFCD00132712
CAS Number: 9050-97-9
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR