Amberlite® XAD-16, Amberlite™ ion exchange resin
Supplier: Thermo Scientific Chemicals
L19565-22
L19565-36
CAAAAL19565-22EA
116.6
CAD
CAAAAL19565-22
CAAAAL19565-36
Amberlite® XAD-16, Amberlite™ ion exchange resin
Amberlite® XAD-16
CAS No.: 104219-63-8
MDL No.: MFCD00145831
Formula:
C₁₀H₁₀ Storage Temperature: Ambient |
MDL Number:
MFCD00145831 CAS Number: 104219-63-8 Merck Index: 14,382 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...