Amberlite XAD-7, Amberlite™ ion exchange resin

Supplier: Thermo Scientific Chemicals

L19564-22 L19564-36
CAAAAL19564-22EA 65 CAD
CAAAAL19564-22 CAAAAL19564-36
Amberlite XAD-7, Amberlite™ ion exchange resin
Amberlite XAD-7

CAS No.: 37380-43-1 MDL No.: MFCD00132705

Storage Temperature: Ambient
MDL Number: MFCD00132705
CAS Number: 37380-43-1
Merck Index: 14,382

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR