4-Chloro-3-(trifluoromethyl)pyridine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19556-03
L19556-MD
CAAAAL19556-03EA
344.44
CAD
CAAAAL19556-03
CAAAAL19556-MD
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride 97%
4-Chloro-3-(trifluoromethyl)pyridine hydrochloride
Purity: 97%
CAS No.: 732306-24-0
Molecular Formula: C6H3ClF3N.HCl
Formula Weight: 218.01
Melting Point: ca 135° subl.
MDL No.: MFCD04972770
Formula:
C₆H₄Cl₂F₃N Melting Pt: ca 135 °C (subl.) Storage Temperature: Ambient |
MDL Number:
MFCD04972770 CAS Number: 732306-24-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...