4-(Trifluoromethyl)nicotinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L19553-03 L19553-MD
CAAAAL19553-03EA 184.86 CAD
CAAAAL19553-03 CAAAAL19553-MD
4-(Trifluoromethyl)nicotinic acid 98%
4-(Trifluoromethyl)nicotinic acid

Purity: 98% CAS No.: 158063-66-2 Molecular Formula: C7H4F3NO2 Formula Weight: 191.11 Melting Point: 147-149° MDL No.: MFCD00082626

Formula: C₇H₄F₃NO₂
MW: 191.11 g/mol
Melting Pt: 147…149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082626
CAS Number: 158063-66-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR